Learn More
Kuhner Shaker Inc SHAKING MACHINE ORBSHAKE SB200
Catalog No. 502569959
Encompass
Shaking machine OrbShake SB200-XTemperature range with cooling: Low: Depends on cooling liquid and culture; High: 50C50mm orbital diameter w/ speeds up to 80rpmControlled acceleration rate includedActive and Positional braking: included and adjustableTimer included and adjustableDC magnetic direct drive with single eccentric (no belt)100% stainless steel interiorDimensions: H2216 x W1013 x D991mm; Weight: 400kgFIVE YEAR WARRANTY HARDWARE
Catalog No. 50-256-9959 Supplier Kuhner Shaker Inc Supplier No. SMX7100
Add to Cart
Add to Cart
Request a Quote
Shaking machine OrbShake SB200-XTemperature range with cooling: Low: Depends on cooling liquid and culture; High: 50C50mm orbital diameter w/ speeds up to 80rpmControlled acceleration rate includedActive and Positional braking: included and adjustableTimer included and adjustableDC magnetic direct drive with single eccentric (no belt)100% stainless steel interiorDimensions: H2216 x W1013 x D991mm; Weight: 400kgFIVE YEAR WARRANTY HARDWARE