Learn More
Cole-Parmer Stuart Shaking Water Bath, 230 V
Supplier:  Cole-Parmer 1215202
Encompass_Preferred
This versatile water bath can be a linear or orbital shaker, or use as a static bath, depending on the platform you choose. Changing platforms is easy — strong magnets couple to the drive unit so there's no need for tricky fittings. The stainless steel shaking platforms come with high-tension springs which accommodate most bottle and flask sizes; with capacity of 8 x 250 mL, 6 x 500 mL, or 4 x 1 L Erlenmeyer flasks. For tubes and smaller vessels, order an optional stainless steel rack. An accessory bath cover is required to reach temperatures above 90°C (order separately).
Analog dials make setting speed and temperature quick, and the digital display ensures your samples will maintain optimum temperature. Stainless steel tank with integral drain makes cleaning and maintenance fast and efficient.
Catalog No. 50-196-0429