Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Shallow Water Outfit
SDPSCI_ED

Catalog No. S85004 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S85004 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S85004 Supplier LaMotte™ Supplier No. 585402
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Turbidity in shallow water is no problem using this standardized reagent.

  • Easily measure turbidity when water is too shallow for a Secchi disk
  • Use a standardized turbidity reagent to match turbidity of a water sample
  • Test four basic water quality factors; all necessary apparatus and reagents included
  • Test factors: Temperature, pH (3.0-10.00, Turbidity Drop Count (0-200 JTU's), Dissolved Oxygen (0-10ppm)

Order Info

Grades 5+.

Specifications

Grade 5+
Product Type Shallow Water Testing Outfit
Includes Necessary apparatus and reagents
For Use With (Application) Easily measure turbidity when water is too shallow for a secchi disk
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.