Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shamrock Labels Biohazard Labels
SDP

Catalog No. 1187310 Shop All Shamrock Scientific Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-873-10 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-873-10 Supplier Shamrock Labels Supplier No. SBH1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides immediate visual warning

  • Brilliant red fluorescent color attracts attention
  • Printed in compliance with OSHA standards for bloodborne pathogens
  • Glove-friendly, permanent adhesives holds labels firmly in place

Specifications

Certifications/Compliance OSHA
Adhesive Type Permanent
Color Red
Format Label
Product Type Label
For Use With (Application) Biohazard
Quantity 500/Pk.
Width (English) 3 in.
Height (English) 2.25 in.
Height (Metric) 5.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.