Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shamrock Labels Formalin Labels
SDP

Catalog No. 1798686g Shop All Shamrock Scientific Products
Encompass
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-986-86G White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-986-86G Supplier Shamrock Labels Supplier No. SB381H
Only null left
Add to Cart
Add to Cart

Specifications

Adhesive Type Permanent
Color White
Format Label
Material Litho
Product Type Label
For Use With (Application) Formalin
Quantity 500/Pk.
Size (English) 1 x 2 in.
Size (Metric) 2.54 x 5.08 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.