Learn More
Epredia™ Shandon™ AN-65 Series Cadaver Lift with Forks, AN-65 Cadaver Lift with Forks, Self-propelled
Catalog No. 881090 Shop All Epredia Products
Change view
Click to view available options
:
Self-propelled
Catalog No.
881090 Self-propelled
1 options
Catalog No. 881090 Supplier Epredia™ Supplier No. 881090
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Minimize physical lifting and handling of cadavers with our heavy-duty lift.
Features:
  • Manual and self-propelled models are available with configurations to manage varying body trays and tasks; also available with spreader bar and strap assembly upon request
  • Forks accommodate standard or bariatric body trays
  • Hydraulic lifting mechanism steadily raises and lowers loads with two forks
  • Raise/lower lever, located between the chassis uprights, provides a fixed lifting speed and variable lowering speed (manual unit) or fixed lowering speed (self-propelled unit)
  • Polyurethane wheels prevent marring of floors
  • Plexiglas™ shield places a barrier between the operator and the chains and cylinder
Ordering Information:
  • Available as self-propelled or manual unit