Learn More
Epredia™ Shandon™ Autopsy Headrests, Rubber
Catalog No. 407 Shop All Epredia Products
Change view
Click to view available options
Description:
Rubber
Catalog No. Description
407 Rubber
1 options
Catalog No. 407 Supplier Epredia™ Supplier No. 407
Add to Cart
Add to Cart
Epredia™ Shandon Autopsy Headrests feature beveled corners to protect autopsy surfaces from scratches.
  • Ten positions of height, width, length and curvature vary on the faces of the headrests for maximum accommodation
  • Only four points contact the table, and base is curved to permit drainage materials to flow freely
  • Each side of the unit is deeply curved to hold the head securely
  • Available in autoclavable aluminum or semi-hard rubber
Specifications
Product Type Autopsy Headrest
Description Rubber