Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Shop All Epredia ProductsDescription
- Writing face is flat 45° angle
- Locking lid attached by two hinges
- Front or back opening
Ordering Information:
- Packaged in 75-cassette tubes
Specifications
Specifications
| Pore Style | Small Square Pore |
| Writing Face Angle | 45° |
| Color | Blue |
| Product Type | Biopsy Cassette |