Learn More
Epredia™ Cryochrome™ Embedding Resin
Catalog No. 9990426 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cryochrome Blue
Cryochrome Green
Cryochrome Pink
Cryochrome Sampler Colors
Cryochrome Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
9990426 Cryochrome Sampler Colors
9990422 Cryochrome Blue
9990423 Cryochrome Yellow
9990428 Cryochrome Green
9990429 Cryochrome Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 9990426 Supplier Epredia™ Supplier No. 9990426
Add to Cart
Edge
Add to Cart
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Product Type Cryochrome Sampler Colors
Quantity 120mL