Learn More
Epredia™ Shandon™ Digital Scale, Replacement bow
Catalog No. 1000232 Shop All Epredia Products
Change view
Click to view available options
Type:
Digital Scale (Order Stand Separately)
Digital Scale Replacement Pan
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
1000232 Digital Scale Replacement Pan
1330 Digital Scale (Order Stand Separately)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1000232 Supplier Epredia™ Supplier No. 1000232
Get accurate autopsy and pathology weight measurements with the Shandon™ Digital Scale.
  • Displays readings with bright, easy-to-read LCD
  • Measures up to 19.99kg
  • Includes ring, bow and pan with drainage holes
  • Battery operated
Compatible with:
Replacement parts may also be used with Shandon™ Analog Scale (Cat. No. 1320).
Specifications
Product Line Shandon
Capacity 19.99 kg
Display Type Digital LCD
Type Digital Scale Replacement Pan