Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs
Catalog No. B1000729YW7 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Peach
Yellow
Quantity:
25 Cassettes x 30 Tubes
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
B1000729YW7 Yellow 25 Cassettes x 30 Tubes
B1000729AQ7 Aqua 25 Cassettes x 30 Tubes
B1000729PE7 Peach 25 Cassettes x 30 Tubes
B1000729LI7 Lilac 25 Cassettes x 30 Tubes
B1000729BL7 Blue 25 Cassettes x 30 Tubes
B1000729GR7 Green 25 Cassettes x 30 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. B1000729YW7 Supplier Epredia™ Supplier No. B1000729YW7
This item is not returnable. View return policy
Compatible with Epredia™ Shandon™ MicroWriter™ cassette labelers
  • Designed to hold tissue specimens during processing and embedding
  • Acetal polymer cassettes with writing face at 45° angle
  • Pore style: Slotted
  • Locking lid attached by two hinges
Specifications
Quantity 25 Cassettes x 30 Tubes
Color Yellow
Material Acetal Polymer