Learn More
Epredia™ Embedding Cassettes in Tube Packs
Catalog No. B1000732AQ Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B1000732AQ Aqua
B1000732BL Blue
B1000732GR Green
B1000732GY Gray
B1000732LI Lilac
B1000732OR Orange
B1000732PE Peach
B1000732PK Pink
B1000732TN Tan
B1000732WH White
B1000732YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. B1000732AQ Supplier Epredia™ Supplier No. B1000732AQ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hold and identify tissue samples in processing, embedding and sectioning procedures with Embedding Cassettes in Tube Packs. Compatible for use with the Epredia™ PrintMate AS™ Cassette Printer.
  • Can be used on all Epredia PrintMate automatic cassette labelers
  • Fit securely in microtome chuck adapters
  • Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
  • Made of acetal polymer with a pore style of round holes
  • Supplied in 10 tubes of 75 cassettes each
Specifications
Pore Style Round Holes
Quantity 750 Cassettes/Cs.
Color Aqua
Material Acetal Polymer