Learn More
Epredia™ EZ Double Cytofunnel™
Catalog No. A78710005 Shop All Epredia Products
Change view
Click to view available options
Quantity:
200/Cs.
40 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A78710005 40 Pk.
A78710021 200/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A78710005 Supplier Epredia™ Supplier No. A78710005
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Double the number of sample presentations per run with the Epredia™ EZ Double Cytofunnel™.
  • Disposable sample chambers with white filter cards and caps
  • Funnel markings corresponding to slide markings for simple identification
  • Pre-attached filter card
Specifications
Color White
Disposable Single-use
Quantity 40 Pk.
Type EZ Double Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.05 to 0.25 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov