Learn More
Epredia™ EZ Megafunnel™ with Caps and Slides
Catalog No. A78710001 Shop All Epredia Products
Change view
Click to view available options
Quantity:
25/Cs.
Catalog No. Quantity
A78710001 25/Cs.
1 options
This item is not returnable. View return policy
Catalog No. A78710001 Supplier Epredia™ Supplier No. A78710001
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prepare larger volume samples conveniently with the Epreida™ EZ Megafunnel™ with Caps and Slides.
  • Minimizes the need to pre-centrifuge
  • Increases cell yield with cell deposition area of 22 x 14.75 mm (325 mm2)
  • Prepares both air-dried and fixed preparations, providing additional valuable diagnostic information
  • Includes Epredia™ EZ Megafunnel™ Cytoslides™
Specifications
Quantity 25/Cs.
Type Megafunnel
WARNING: Reproductive Harm - www.P65Warnings.ca.gov