Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin

Catalog No. 9990914 Shop All Epredia Products
Change view
Click to view available options
Quantity:
120 mL container 60 mL filled
3.8 L
5 gal. (19L)
60 mL container 30 mL filled
90 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9990914 60 mL container 30 mL filled
6764240 5 gal. (19L)
6764254 3.8 L
9990244 -
9990916 90 mL
9990918 120 mL container 60 mL filled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 9990914 Supplier Epredia™ Supplier No. 9990914
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.

  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available

Specifications

Packaging Type 125/Cs.
Product Type Formalin
Quantity 60 mL container 30 mL filled
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.