Description
- Phosphate-free
- Use for storage of all tissue types
- Stores specimens without excessive shrinking or hardening
- Available premixed, ready-to-use, or in economical concentrate
- Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
- Pre-filled, leak-resistant cups are available
Specifications
Specifications
| Packaging Type | 125/Cs. |
| Product Type | Formalin |
| Quantity | 120 mL container 60 mL filled |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov