Learn More
Epredia™ Shandon™ Frozen Section Staining Kit, Support Rack
Catalog No. 23145 Shop All Epredia Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
23-145 Stainless Steel
1 options
Catalog No. 23-145 Supplier Epredia™ Supplier No. 23145
Add to Cart
Add to Cart
For use with the Epredia™ Shandon™ Frozen Section Staining Kit.
  • Stainless-steel support rack measures 3.5 x 4.5 x 17.25 in. when rack is closed
Specifications
Material Stainless Steel
Product Type Support Rack