Learn More
Epredia™ Glyo-Fixx™
Catalog No. 6764260 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
125 x 60mL
180 x 20mL
5 gal.
125 x 120mL
Product Type:
Fixative
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
6764260 5 gal. Fixative
67-642-62 1 gal. Fixative
9990924 125 x 60mL Fixative
9990928 125 x 120mL Fixative
9990920 180 x 20mL Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6764260 Supplier Epredia™ Supplier No. 6764260
Add to Cart
Add to Cart
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
Specifications
Product Type Fixative
Quantity 5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov