Learn More
Epredia™ Glyo-Fixx™
Catalog No. p-4532387
Click to view available options
Quantity:
1 gal.
125 x 60mL
180 x 20mL
5 gal.
125 x 120mL
Product Type:
Fixative
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
Specifications
Product Type Fixative
Quantity 5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.