Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Instant Eosin

Catalog No. p-4532352 Shop All Epredia Products
Change view
Click to view available options
Solution Type:
Eosin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Solution Type
6765040 Eosin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6765040 Supplier Epredia™ Supplier No. 6765040
Only null left
Add to Cart
Add to Cart

Obtain brilliant and consistent staining with Epredia™ Shandon™ Instant Eosin.

  • Stable and precipitate-free; no filtering
  • pH-adjusted for maximum stain uptake
  • Economical powder stores indefinitely
  • Compatible with all alum Hematoxylin stains

Specifications

Compatibility Alum Hematoxylin Stains
Product Type Eosin
Solution Type Eosin
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.