Learn More
Epredia™ Shandon™ Instant Hematoxylin
Catalog No. 6765015 Shop All Epredia Products
Change view
Click to view available options
Quantity:
Makes 1L x 6; 6/pack
Catalog No. Quantity
6765015 Makes 1L x 6; 6/pack
1 options
This item is not returnable. View return policy
Catalog No. 6765015 Supplier Epredia™ Supplier No. 6765015
Add to Cart
Add to Cart
This item is not returnable. View return policy
Substitute Epredia™ Shandon™ Instant Hematoxylin for Harris or Gill when no mucoid staining is required.
  • Stains from blue-black to light blue depending upon preparation
Specifications
Quantity Makes 1L x 6; 6/pack
Product Type Instant Hematoxylin