Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Kwik-Diff™ Stains

Catalog No. 9990706 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
3 x 500 mL
4 L
Product Type:
Eosin
Fixative
Kwik-Diff Stains
Methylene Blue
Includes:
125 mL Each: Fixative, Eosin, Methylene Blue
4 L Kwik-Diff Reagent 1, Fixative
4 L Kwik-Diff Reagent 2, Eosin
4 L Kwik-Diff Reagent 3, Methylene Blue
500 mL Each: Fixative, Eosin, Methylene Blue
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Includes
99-907-06 4 L Eosin 4 L Kwik-Diff Reagent 2, Eosin
99-907-00 3 x 500 mL Kwik-Diff Stains 500 mL Each: Fixative, Eosin, Methylene Blue
9990705 4 L Fixative 4 L Kwik-Diff Reagent 1, Fixative
99-907-07 4 L Methylene Blue 4 L Kwik-Diff Reagent 3, Methylene Blue
99-907-01 125 mL Kwik-Diff Stains 125 mL Each: Fixative, Eosin, Methylene Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 99-907-06 Supplier Epredia™ Supplier No. 9990706
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.

  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori

Includes:

Reagent 2: Eosin

Specifications

Quantity 4 L
Includes 4 L Kwik-Diff Reagent 2, Eosin
Product Type Eosin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.