Learn More
Thermo Scientific™ Shandon™ Nu-Clear™ Available on GSA/VA Contract for Federal Government customers only.
Achieve improved cell clarity by removing excess hematoxylin from the slide with Thermo Scientific™ Shandon™ Nu-Clear™, a specially-formulated acid alcohol solution.
$92.10 - $121.10
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
6769008 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
6769008
Nu-Clear I
Each for $121.10
Add to cart
6769009 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
6769009
Nu-Clear II
Each for $92.10
Add to cart
Description
Description
Achieve improved cell clarity by removing excess hematoxylin from the slide with Thermo Scientific™ Shandon™ Nu-Clear™, a specially-formulated acid alcohol solution.
Key Features
  • Removes residual hematoxylin staining the glass.
  • Designed to be used on the full line of Thermo Scientific Shandon automatic stainers as well as manual methods.
Nu-Clear I
  • Fast-acting solution
  • Recommended to be used as a 10-second step in most protocols
  • Particularly beneficial for use with Harris hematoxylin
  • May be used on the Thermo Scientific Varistain™ 24-4 or Shandon Varistain Gemini slide stainers
Nu-Clear II
  • Less concentrated
  • Recommended as a 20-second step
  • May be used on the Thermo Scientific Shandon Linistat and other linear stainers
  • Rcommended for use with Gill hematoxylins
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.