Learn More
Epredia™ Shandon™ Polysine Slides, White
GSA_VA
Catalog No. 6776215 Shop All Epredia Products
Change view
Click to view available options
Description:
Blue; 144/Pack
White; 144/Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
67-762-15 White; 144/Pack
67-762-16 Blue; 144/Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-762-15 Supplier Epredia™ Supplier No. 6776215
Add to Cart
Add to Cart
This item is not returnable. View return policy
Achieve superior cell and tissue adhesion with Epredia™ Shandon™ Polysine Slides.
  • Polysine places an advanced permanent adhesive on the microscope slide
  • Built-in chemicals electrostatically and chemically attract tissues and cells
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Microscope Slide
Material Polysine
Color White
Description White; 144/Pack