Learn More
Epredia™ Shandon™ Polysine Slides, White
Catalog No. 6776216 Shop All Epredia Products
Change view
Click to view available options
Description:
Blue; 144/Pack
White; 144/Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
67-762-16 Blue; 144/Pack
67-762-15 White; 144/Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-762-16 Supplier Epredia™ Supplier No. 6776216
Add to Cart
Add to Cart
This item is not returnable. View return policy
Achieve superior cell and tissue adhesion with Epredia™ Shandon™ Polysine Slides.
  • Polysine places an advanced permanent adhesive on the microscope slide
  • Built-in chemicals electrostatically and chemically attract tissues and cells
Specifications
Type Microscope Slide
Material Polysine
Color Blue
Description Blue; 144/Pack
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov