Learn More
Epredia™ Rapid-Fixx™ Formalin Fixative
Catalog No. 6764212 Shop All Epredia Products
Change view
Click to view available options
Quantity:
2 x 1 L
Catalog No. Quantity
6764212 2 x 1 L
1 options
This item is not returnable. View return policy
Catalog No. 6764212 Supplier Epredia™ Supplier No. 6764212
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Obtain quick results with this formalin. Excellent for antigenic preservation, avoids the need for enzyme digestion, and rapidly penetrates into tissue.
Specifications
Quantity 2 x 1 L
What fixative should I use to fix my cells or tissues for antibody labeling?
Aldehyde-based fixatives (e.g., formaldehyde, glutaraldehyde) crosslink various cellular components, which helps retain proteins and cell morphology, but some antigens can be masked by the crosslinking, requiring antigen retrieval methods to unmask the antigen binding sites. Also, aldehyde-based fixation requires permeabilization to allow entry of antibodies. Organic solvents such as methanol and acetone condense proteins and permeabilize the cells, but cell morphology can be compromised. Surface antigens can be labeled without fixation.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov