Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Slide Holder, 10 Slide Capacity

Catalog No. 147 Shop All Epredia Products
Change view
Click to view available options
Holds:
Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
14-7 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-7 Supplier Epredia™ Supplier No. 147
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Store 10 slides vertically with the Epredia™ Shandon™ Slide Holder.

  • Features individually stamped slots
  • Holds 76 x 26mm

Specifications

Holds Slides
Positions 10
Length (Metric) Slide 76 mm
Width (Metric) Slide 26 mm
Product Type Slide Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.