Learn More
Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades
Catalog No. 5334 Shop All Epredia Products
Change view
Click to view available options
Size:
# 4
Catalog No. Size
53-34 # 4
1 options
This item is not returnable. View return policy
Catalog No. 53-34 Supplier Epredia™ Supplier No. 5334
Add to Cart
Add to Cart
This item is not returnable. View return policy
Securely hold blades with the Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, constructed of #4 stainless steel.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
Specifications
Blade Material Stainless Steel
Size # 4
Length (English) 8.5 in.
Length (Metric) 21.6 cm
For Use With (Equipment) Holds # 20 to # 25 Blade