Learn More
Epredia™ TBD-1™ Decalcifier
Catalog No. p-4532362 Shop All Epredia Products
Change view
Click to view available options
Quantity:
2 Pk.
Catalog No. Quantity
67-640-01 2 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 67-640-01 Supplier Epredia™ Supplier No. 6764001
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Process bone marrow biopsies in as little as 15 to 30 minutes with this rapid hydrochloric acid decalcifier.
  • Process thin pieces of dense bone in 3 to 12 hours
  • Permits utilization of routine end point tests, such as calcium oxalate
  • Protects cells from actions of acids with added ingredients
Specifications
Product Type Decalcifying Solution
Quantity 2 Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov