Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Catalog No. p-4524749 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
10-009-57 White
10-009-59 Green
10-009-60 Yellow
10-009-61 Blue
10-009-62 Peach
10-009-64 Gray
10-009-65 Lilac
10-009-66 Orange
10-009-67 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 10-009-57 Supplier Epredia™ Supplier No. 1000957
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
  • Acetal polymer construction
  • 30° writing face
  • Small slot-style pores
Ordering Alerts:
  • Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Pore Style Slotted Pore
Writing Face Angle 30°
Color White
Material Acetal Polymer
Product Type Tissue Cassette