Learn More
Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Catalog No. 22024964 Shop All Epredia Products
Encompass
Change view
Click to view available options
Quantity:
59 mL
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
22-024-964 59 mL Green
NC9513663 59 mL Blue
NC9601922 59 mL Yellow
22-024-963 59 mL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-024-964 Supplier Epredia™ Supplier No. 3120128
Add to Cart
Add to Cart
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Color Green
Product Type Tissue Marking Dye
Quantity 59 mL