Learn More
Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Catalog No. NC9513663 Shop All Epredia Products
Encompass
Change view
Click to view available options
Quantity:
59 mL
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
NC9513663 59 mL Blue
NC9601922 59 mL Yellow
NC9513665 59 mL Red
NC9513664 59 mL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NC9513663 Supplier Epredia™ Supplier No. 3120124
Only null left
Add to Cart
Add to Cart
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Color Blue
Product Type Tissue Marking Dye
Quantity 59 mL