Learn More
Epredia™ Shandon™ Wright-Giemsa Stain Kit
Catalog No. 9990710 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 mL
Product Type:
Wright-Giemsa Stain
Catalog No. Quantity Product Type
99-907-10 500 mL Wright-Giemsa Stain
1 options
Catalog No. 99-907-10 Supplier Epredia™ Supplier No. 9990710
Add to Cart
Add to Cart
Eosinates of polychrome methylene blue yield rich color
  • Unique manufacturing process produces an intense differential stain for blood smears, bone marrows, mast cells, and FNA's
  • Demonstrates microorganisms such as Microfilaria, Babesia, Haemobartonella, Giardia, Pneumocystis, and Helicobacter
Specifications
Quantity 500 mL
Includes Stain, buffer and rinse
Product Type Wright-Giemsa Stain
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov