Learn More
TARGETMOL CHEMICALS INC Shanzhiside methyl ester 25MG
SDP
Catalog No. 502879184
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Shanzhiside methylester reduces neuropathic pain by activating spinal GLP-1 receptors and subsequently stimulating microglial beta-endorphin expression via the p38 MAPK signaling. Purity 99.32%
Catalog No. 50-287-9184 Supplier TARGETMOL CHEMICALS INC Supplier No. T389625MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Shanzhiside methylester reduces neuropathic pain by activating spinal GLP-1 receptors and subsequently stimulating microglial beta-endorphin expression via the p38 MAPK signaling. Purity 99.32%