Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Shanziside | 29836-27-9 | 20 MG
SDP

Catalog No. 5000418771
Encompass_Preferred

Shanziside is an iridoid glucoside isolated from Phlomis tuberosa L. This compound is for research use only. It appears as a solid, white to off-white in color, with a molecular weight of 392.36 and a chemical formula of C16H24O11. It is classified under terpenoids and iridoids, originating from plants like Leguminosae (Phaseolus lunatus L.) and Labiatae.

  • Isolated from Phlomis tuberosa L.
  • Intended for research use only
  • Solid, white to off-white appearance
  • Molecular weight of 392.36
  • Chemical formula C16H24O11
  • Classified as an iridoid glucoside, terpenoid
  • Soluble in DMSO at 50 mg/mL (requires ultrasonic assistance)
  • Store at -20°C, sealed, away from moisture and light; in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-18771 Supplier Medchemexpress LLC Supplier No. HYN409220MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Shanziside is an iridoid glucoside isolated from Phlomis tuberosa L. This compound is for research use only. It appears as a solid, white to off-white in color, with a molecular weight of 392.36 and a chemical formula of C16H24O11. It is classified under terpenoids and iridoids, originating from plants like Leguminosae (Phaseolus lunatus L.) and Labiatae.

  • Isolated from Phlomis tuberosa L.
  • Intended for research use only
  • Solid, white to off-white appearance
  • Molecular weight of 392.36
  • Chemical formula C16H24O11
  • Classified as an iridoid glucoside, terpenoid
  • Soluble in DMSO at 50 mg/mL (requires ultrasonic assistance)
  • Store at -20°C, sealed, away from moisture and light; in solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.