Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Shanziside | 29836-27-9 | 99.9% | 392.36 | C16H24O11 | 10 MG
SDP

Catalog No. 5000289802
Encompass_Preferred

Shanziside is an iridoid glucoside isolated from Phlomis tuberosa L., supplied for research use. The compound has molecular formula C16H24O11, molecular weight 392.36, and CAS number 29836-27-9. It is typically provided in small milligram quantities for biochemical and pharmacological studies, with established solubility and storage recommendations for experimental handling.

  • Iridoid glucoside used in biochemical research.
  • High purity: 99.87%.
  • Molecular formula C16H24O11; molecular weight 392.36.
  • Soluble in DMSO at 50 mg/mL with ultrasonic assistance.
  • Store at -20°C, sealed, away from moisture and light; in solution store at -80°C for long-term stability.
  • Supplied as a 10 MG vial suitable for small-scale studies.

Catalog No. 50-002-89802 Supplier Medchemexpress LLC Supplier No. HYN409210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Shanziside is an iridoid glucoside isolated from Phlomis tuberosa L., supplied for research use. The compound has molecular formula C16H24O11, molecular weight 392.36, and CAS number 29836-27-9. It is typically provided in small milligram quantities for biochemical and pharmacological studies, with established solubility and storage recommendations for experimental handling.

  • Iridoid glucoside used in biochemical research.
  • High purity: 99.87%.
  • Molecular formula C16H24O11; molecular weight 392.36.
  • Soluble in DMSO at 50 mg/mL with ultrasonic assistance.
  • Store at -20°C, sealed, away from moisture and light; in solution store at -80°C for long-term stability.
  • Supplied as a 10 MG vial suitable for small-scale studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.