Learn More
Sielc Technologies Inc SHARC 1 1MM DIA 150MM LENGTH
Catalog No. NC3993808
Encompass
SHARC™ columns represent the first commercially available columns that primarily utilize separation based on hydrogen bonding. SHARC™ is an acronym for Specific Hydrogen-bond Adsorption Resolution Chromatography.Column Diameter 1 mmColumn Length 150 mmParticle Size 3 µmPore Size 100 AColumn options Surface coatedClear
Catalog No. NC3993808 Supplier Sielc Technologies Inc Supplier No. SH110.150.0310.C
SHARC™ columns represent the first commercially available columns that primarily utilize separation based on hydrogen bonding. SHARC™ is an acronym for Specific Hydrogen-bond Adsorption Resolution Chromatography.Column Diameter 1 mmColumn Length 150 mmParticle Size 3 µmPore Size 100 AColumn options Surface coatedClear