Learn More
Cayman Chemical Squalene, Purity: ≥95%, CAS: 111-02-4, 100 g
SDP
Catalog No. 502937784
Encompass_Preferred
A biosynthetic precursor to steroids; an oil-in-water emulsion synergistically increases adaptive immune responses to GLA compared with an aqueous formulation of GLA
Catalog No. 50-293-7784 Supplier Cayman Chemical Supplier No. 27058100 G
A biosynthetic precursor to steroids; an oil-in-water emulsion synergistically increases adaptive immune responses to GLA compared with an aqueous formulation of GLA