Learn More
Cayman Chemical Squalene, Purity: ≥95%, CAS: 111-02-4, 50 g
SDP
Catalog No. 502939837
Encompass_Preferred
A biosynthetic precursor to steroids; an oil-in-water emulsion synergistically increases adaptive immune responses to GLA compared with an aqueous formulation of GLA
Catalog No. 50-293-9837 Supplier Cayman Chemical Supplier No. 2705850 G
A biosynthetic precursor to steroids; an oil-in-water emulsion synergistically increases adaptive immune responses to GLA compared with an aqueous formulation of GLA