Learn More
International Medical Industries (IMI) Aspirating Needle Sharp Cannula 19 Gauge X 5 Inch (Box of 100)
Catalog No. NC2030217
Encompass
Rx-Tract™ Aspirating Needle provides high-quality solutions for introducing fluids to deep wells and vials, drawing liquids for reconstruction, or pre-fill syringes prior to administration. Rx-Tract Aspirating Needles comes with a conveniently sealed guard to enhance pharmacist and clinician safety. •Stainless Steel Needle • Sterile • Non-Pyrogenic • Individual Blister • Box of 100 • 19 Gauge • 5 Inch (7.62 CM) • Sharp Cannuala
Catalog No. NC2030217 Supplier International Medical Industries (IMI) Supplier No. 3222
Rx-Tract™ Aspirating Needle provides high-quality solutions for introducing fluids to deep wells and vials, drawing liquids for reconstruction, or pre-fill syringes prior to administration. Rx-Tract Aspirating Needles comes with a conveniently sealed guard to enhance pharmacist and clinician safety. •Stainless Steel Needle • Sterile • Non-Pyrogenic • Individual Blister • Box of 100 • 19 Gauge • 5 Inch (7.62 CM) • Sharp Cannuala