Learn More
Grainger Young Brothers Stamp Works Steel Type Kit, Round-Face, 1/8 in Character Height, 106 Pieces
Catalog No. 502787985
Encompass_Preferred
This steel type kit is designed for precise and aligned marking of serials, dates, and part codes in a single stroke. The set features durable steel construction with a round-face character style, suitable for materials with a surface hardness up to 50 HRC.
  • Character height of 1/8 in.
  • Includes letters A to Z, numbers 0 to 8 (with 6 serving as 9), an ampersand, slash, period, comma, spacers, and dash.
  • Steel shank length of 7/8 in.
  • Maximum capacity of 8 characters per line.
  • Includes a type holder and organized plastic storage case.
  • Rockwell hardness of 50 on the C scale.
Catalog No. 50-278-7985 Supplier Grainger Supplier No. 18G263
This steel type kit is designed for precise and aligned marking of serials, dates, and part codes in a single stroke. The set features durable steel construction with a round-face character style, suitable for materials with a surface hardness up to 50 HRC.
  • Character height of 1/8 in.
  • Includes letters A to Z, numbers 0 to 8 (with 6 serving as 9), an ampersand, slash, period, comma, spacers, and dash.
  • Steel shank length of 7/8 in.
  • Maximum capacity of 8 characters per line.
  • Includes a type holder and organized plastic storage case.
  • Rockwell hardness of 50 on the C scale.