Learn More
Electron Microscopy Sciences Sharp Objects Dispo-Pouch
SDP
Catalog No. 5028597
Encompass_Preferred
Made of 10 mil solid paperboard, which opens to an oval shape by folding the bottom flaps. When full just close the top flaps and lock into place. Ideal for needles, razor blades, scalpel blades, etc.
Catalog No. 50-285-97 Supplier Electron Microscopy Sciences Supplier No. 64431
Made of 10 mil solid paperboard, which opens to an oval shape by folding the bottom flaps. When full just close the top flaps and lock into place. Ideal for needles, razor blades, scalpel blades, etc.