Learn More
Electron Microscopy Sciences Sharp Objects Dispo-Pouch,
SDP
Catalog No. 5028599
Encompass_Preferred
Made of 10 mil solid paperboard, which opens to an oval shape by folding the bottom flaps. When full just close the top flaps and lock into place. Ideal for needles, razor blades, scalpel blades, etc.
Catalog No. 50-285-99 Supplier Electron Microscopy Sciences Supplier No. 64430
Made of 10 mil solid paperboard, which opens to an oval shape by folding the bottom flaps. When full just close the top flaps and lock into place. Ideal for needles, razor blades, scalpel blades, etc.