Learn More
Sklar Instruments Sharpening Stone, Sklar, Sharpen the tips of Keyes Biopsy Punch. 5-1/2 in. EA.
SDP
Catalog No. 501183563 Shop All Sklar Instruments Products
Encompass_Preferred
Sharpening Stone, Sklar, Sklar's Sharpening Stone for Keyes Biopsy Punches is a stainless steel and diamond pen-like tool used to sharpen the tips of Keyes Biopsy Punch. 5-1/2 in.
Catalog No. 50-118-3563 Supplier Sklar Instruments Supplier No. 064110
Sharpening Stone, Sklar, Sklar's Sharpening Stone for Keyes Biopsy Punches is a stainless steel and diamond pen-like tool used to sharpen the tips of Keyes Biopsy Punch. 5-1/2 in.