Learn More
Fine Science Tools Sharpening Stones, 6 mm x 9 cm, each
SDP
Catalog No. NC9681212
Encompass
Natural Arkansas oil stones feature unequalled fineness, hardness and structural density. When used with instrument oil, they will produce the sharpest cutting edge. Square, 6mmx9cm.
Catalog No. NC9681212 Supplier Fine Science Tools Supplier No. 2900801
Natural Arkansas oil stones feature unequalled fineness, hardness and structural density. When used with instrument oil, they will produce the sharpest cutting edge. Square, 6mmx9cm.