Learn More
Electron Microscopy Sciences Pre-sharpened Single Tip Carbon Rod, 2-3/8" OAL, ⅛" diameter
SDP
Catalog No. 5024910
Encompass_Preferred
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
OAL: 2-3/8"
Diameter: ⅛"
Tip diameter: 0.205”
Tip: 0.205” neck
Catalog No. 50-249-10 Supplier Electron Microscopy Sciences Supplier No. 7022001
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
OAL: 2-3/8"
Diameter: ⅛"
Tip diameter: 0.205”
Tip: 0.205” neck