Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sharps-A-Gator™ Point-of-Use Sharps Containers

Catalog No. 14827129 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
5 qt.
3.5 gal.
Color:
Red
Translucent Red
Dimensions (L x W x H):
4.25 x 11 x 12.25 in.
8 x 14.25 x 13.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color Dimensions (L x W x H)
14-827-129 3.5 gal. Translucent Red 8 x 14.25 x 13.5 in.
14-827-122 5 qt. Red 4.25 x 11 x 12.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-827-129

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Horizontal loading, nonmechanical sharps opening provides one-hand, one-step, no-touch sharps depositing.

 

  • For use in patient rooms and all patient-care areas Opening design impedes hand entry to reduce risk of needle sticks when disposing of sharps
  • Clear tops and translucent containers allow you to monitor fill level
  • Puncture-resistant, durable polypropylene 
  • Clearly marked with biohazard label
  • Non-Autoclavable

 

Specifications

Capacity (Metric) 13.2 L
Closure Type Dual Opening Hinge w/ VAT Perimeter Locks, Temporary
Capacity (English) 3.5 gal.
Color Translucent Red
Dimensions (L x W x H) 8 x 14.25 x 13.5 in.
Disposable Single-use
Height (Metric) 34.3 cm
Length (Metric) 20.3 cm
Quantity 10/Cs.
Width (Metric) 36 cm
Autoclavable Not Autoclavable
Width (English) 14.25 in.
Length (English) 8 in.
Height (English) 13.5 in.
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.