Learn More
Heathrow Scientific Sharps Bank Biomedical Waste Disposal Unit, 15/Cs.
Description
- Sturdy Corrugated Construction
- Up to 70% less expensive than plastic containers
- Handle design and location promotes safe handling
- Square, flat bottom reduces the chance of tipping
- Three-walled construction provides puncture-resistant containment of contaminated waste
- 3 mil liner provides leak resistance
- Extra wide opening for safe, convenient disposal of contaminated waste
- Two flap configuration protects contents from spilling
- Ships and stores flat for space-saving storage
- Made in the USA using recycled materials
Specifications
Specifications
| Capacity (English) | 13 gal. |
| Color | White |
| Dimensions (L x W x H) | 13.25 x 13.25 x 17.35 in. (33.7 x 33.7 x 44.1 cm) |
| Quantity | 15/Cs. |
| Autoclavable | Not Autoclavable |
| Material | Corrugated Cardboard |
| Product Type | Biohazard Container |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.