Learn More
Med Vet International Sharps Container, 1 Gallon, 10/Box
Catalog No. 501180487
Encompass_Preferred
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy-to-assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs.
Catalog No. 50-118-0487 Supplier Med Vet International Supplier No. SHARP1GX10
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy-to-assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs.