Learn More
Sigma Aldrich Fine Chemicals Biosciences Sharps container 1 lb red
Catalog No. 502901491
Encompass
The Sharps Container is easy to use and transport. Provides safe disposal of syringes needles and other small sharps.Stackable to reduce storage space before use1.0 L capacity (Quart) with a clear lid to view fill levelSlide access lid for temporary and final closures for safety and securityBuilt-in needle removal port preventing direct contact with sharpsCompatible with the Droplet Blood Collection TrayMeasures 4.5 x 4.5 x 7.5 in (11.5 x 11.5 x 19.2 cm)Pack of 18
Catalog No. 50-290-1491 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120177
The Sharps Container is easy to use and transport. Provides safe disposal of syringes needles and other small sharps.Stackable to reduce storage space before use1.0 L capacity (Quart) with a clear lid to view fill levelSlide access lid for temporary and final closures for safety and securityBuilt-in needle removal port preventing direct contact with sharpsCompatible with the Droplet Blood Collection TrayMeasures 4.5 x 4.5 x 7.5 in (11.5 x 11.5 x 19.2 cm)Pack of 18